Learn More
Sino Biological Recombinant Human CLEC4D Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11485H07H100
Encompass_Preferred
A DNA sequence encoding the human CLEC4D (NP_525126.2) extracellular domain (Gly 52-Asn 215) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6909