Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1719786
Encompass_Preferred
Aluminum housed mantle for 1L spherical flask, to 600 degrees C maximum internal operating temperature, 600W, 115V CSA
Catalog No. NC1719786 Supplier Glas Col LLC Supplier No. 100B STM1000
Aluminum housed mantle for 1L spherical flask, to 600 degrees C maximum internal operating temperature, 600W, 115V CSA