Learn More
Sino Biological Recombinant Human Factor XIII Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11510H08H50
Encompass_Preferred
A DNA sequence encoding the human F13B (P05160) (Met 1-Thr 661) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6918