Learn More
Sino Biological Recombinant Human Fibromodulin Protein (hFc Tag)
Supplier:  Sino Biological 11514H02H100
Encompass_Preferred
A DNA sequence encoding the human FMOD (NP_002014.2) (Met1-Ile376) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6921