Learn More
Sino Biological Recombinant Human PTPN9 Protein (His & GST Tag)
Supplier:  Sino Biological 11517H20B100
Encompass_Preferred
A DNA sequence encoding the human PTPN9 (NP_002824.1) (Ala285-Gln593) was expressed with a C-terminal polyhistidine-tagged GST tag at the N-terminus (his-GST).
Catalog No. 50-161-6923