Learn More
Sino Biological Recombinant Human METTL1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11525H07E100
Encompass_Preferred
A DNA sequence encoding the human METTL1 (NP_005362.3) (Asp 32-Gln 265) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-6931