Learn More
Sino Biological Recombinant Human FGF6 Protein
Supplier:  Sino Biological 11528HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human FGF6 (NP_066276.2)(Gly41-Ile208) was expressed with a N-terminal Met.
Catalog No. 50-161-6936