Learn More
Sino Biological Recombinant Human PAK3 Protein (His Tag)
Supplier:  Sino Biological 11532H08B50
Encompass_Preferred
A DNA sequence encoding the human PAK3 isoform 2 (O75914-2) (Met 1-Arg 544) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6941