Learn More
Sino Biological Recombinant Human MST1 Protein (His Tag)
Supplier:  Sino Biological 11533H07B50
Encompass_Preferred
A DNA sequence encoding the human STK4 isoform 1 (Q13043-1) (Glu 2-Phe 487) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6943