Learn More
Sino Biological Recombinant Human NEK7 Protein (His & GST Tag)
Supplier:  Sino Biological 11534H20B50
Encompass_Preferred
A DNA sequence encoding the human NEK7 (NP_598001.1) (Met1-Ser 302) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-6945