Learn More
Sino Biological Recombinant Human CDC42BPB Protein (His & GST Tag)
Supplier:  Sino Biological 11540H20B50
Encompass_Preferred
A DNA sequence encoding the amino acid (Met 1-His 427) of human CDC42BPB (Q9Y5S2) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-6949