Learn More
Sino Biological Recombinant Human TLK2 Protein (aa 397-772, His & GST Tag)
Supplier:  Sino Biological 11551H20B50
Encompass_Preferred
A DNA sequence encoding the amino acids (Leu 397-Asn 772) of human TLK2 (Q86UE8-1) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-6959