Learn More
Sino Biological Recombinant Human DOT1L Protein
Supplier:  Sino Biological 11552HNCE50
Encompass_Preferred
A DNA sequence encoding the human DOT1L (NP_115871.1) N-terminal segment (Gly 2-Lys 416) was expressed and purified, with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-6963