Learn More
Sino Biological Recombinant Human Enolase 1/ENO1 Protein (His Tag)
Supplier:  Sino Biological 11554H07E100
Encompass_Preferred
A DNA sequence encoding the human ENO1 (P06733-1) (Met 1-Lys 434) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-6964