Learn More
Sino Biological Recombinant Human VRK1 Protein (His & GST Tag)
Supplier:  Sino Biological 11555H20B50
Encompass_Preferred
A DNA sequence encoding the human VRK1 (Q99986) (Met 1-Lys 396) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-6967