Learn More
Sino Biological Recombinant Human PTPN12 Protein (aa 1-355, His & GST Tag)
Supplier:  Sino Biological 11556H20B100
Encompass_Preferred
A DNA sequence encoding the human PTPN12 (AAA36529.1) (Met1-Gln355) was expressed with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-6970