Learn More
Sino Biological Recombinant Human PTPN12 Protein
Supplier:  Sino Biological 11556HNCB100
Encompass_Preferred
A DNA sequence encoding the human PTPN12 (AAA36529.1) (Met1-Gln355) was expressed and purified with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-6972