Learn More
Sino Biological Recombinant Human B7-H2/ICOSLG Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11559H08H100
Encompass_Preferred
A DNA sequence encoding the human ICOSLG (NP_056074.1) extracellular domain (Met 1-Ser 258) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6982