Learn More
Sino Biological Recombinant Human NETO1 Protein (His Tag)
Supplier:  Sino Biological 11562H08H100
Encompass_Preferred
A DNA sequence encoding the human NETO1 isoform 3 (NP_620416.1) extracellular domain (Met 1-Thr 344) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6985