Learn More
Sino Biological Recombinant Human Cystatin SN Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11568H08H20
Encompass_Preferred
A DNA sequence encoding the human CST1 (P01037-1) (Met 1-Ser 141) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6990