Learn More
Sino Biological Recombinant Human NLK/Nemo Like Kinase Protein (His & GST Tag)
Supplier:  Sino Biological 11573H20B50
Encompass_Preferred
A DNA sequence encoding the human NLK (Q9UBE8) (Val121-Glu527) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-6994