Learn More
Sino Biological Recombinant Human EDAR Protein (His Tag)
Supplier:  Sino Biological 11577H08H100
Encompass_Preferred
A DNA sequence encoding the human EDAR (NP_071731.1) extracellular domain (Met 1-Ile 189) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7001