Learn More
Sino Biological Recombinant E. coli BirA Protein (His & MBP Tag)
Supplier:  Sino Biological 11582E10E100
Encompass_Preferred
A DNA sequence encoding the E.coil BirA (P06709) (Met1–Lys321) was fused with an N-terminal polyhistidine-tagged MBP tag at the N-terminus.
Catalog No. 50-161-7005