Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Set of Eight Precision Specific Gravity Hydrometers
SDP

Catalog No. 11582 Shop All Fisherbrand Products
Change view
Click to view available options
Certifications/Compliance:
3-point Cert, NIST, ASTM E100, E126
NIST, ASTM E100, ASTM E126
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
11-582 NIST, ASTM E100, ASTM E126
11-582C 3-point Cert, NIST, ASTM E100, E126
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste.
Catalog No. 11-582
Add to Cart
Add to Cart
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste.

Specific Gravity Hydrometers are permanently marked with individual serial numbers.

Set includes thermometer with 0° to 220°F range and eight precision hydrometers that cover a specific gravity range of 0.695 to 2.000.

  • Approximately 300mm long for use with 08-530J cylinder, available separately
  • In lined case

Order Info

A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.

Certifications

A certificate is provided to indicate instrument traceability to standards provided by NIST™.

Compliance

Accurate to one graduation line that meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM™ E 126 specifications.

TRUSTED_SUSTAINABILITY

Specifications

Increments 0.001/0.002
Certifications/Compliance NIST, ASTM E100, ASTM E126
Length (Metric) 300 mm
For Use With (Equipment) 300mL Borosilicate Glass cylinder
Material Soda Lime Glass
Temperature Range (English) 0°F to 220°F
Baume Range 0.695 to 2
For Use With (Application) Specific Gravity Measurement
Specific Gravity Range 0.695 to 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.