Learn More
Sino Biological Recombinant Human LCN1 Protein (His Tag)
Supplier:  Sino Biological 11583H08H100
Encompass_Preferred
A DNA sequence encoding the human LCN1 (NP_002288.1) extracellular domain (Met 1-Asp176) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7007