Learn More
Sino Biological Recombinant Human Placental Lactogen/CSH1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11596H08H50
Encompass_Preferred
A DNA sequence encoding the human CSH1 (NP_001308.1) (Met 1-Phe 217) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7022