Learn More
Sino Biological Recombinant Human STK40 Protein (His & GST Tag)
Supplier:  Sino Biological 11597H20B50
Encompass_Preferred
A DNA sequence encoding the human STK40 (NP_114406) (Met1-Lys435) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7023