Learn More
Sino Biological Recombinant Human ATF2 Protein (His & GST Tag)
Supplier:  Sino Biological 11599H20B100
Encompass_Preferred
A DNA sequence encoding full length of human ATF2 (P15336-1) (Met 1-Ser 505) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7024