Learn More
Labtech Inc Recirculating Water Chille
Catalog No. 501959345
Encompass_Preferred
Recirculating Water Chiller by LabTech, cooling capacity of 1000 Watt at 25˚C, 115V, 8C ~ 35˚C, model H150-1000-115
Catalog No. 50-195-9345 Supplier Labtech Inc Supplier No. LW1000115V
Add to Cart
Add to Cart
Recirculating Water Chiller by LabTech, cooling capacity of 1000 Watt at 25˚C, 115V, 8C ~ 35˚C, model H150-1000-115