Learn More
Perkin Elmer US LLC 1/16, 10-Port Gas Sample Valve with T rotor
Catalog No. 502104023
Encompass_Preferred
The valve has a maximum temperature of 350°C and a maximum pressure of 300 psi.
Catalog No. 50-210-4023 Supplier Perkin Elmer US LLC Supplier No. N9302816
The valve has a maximum temperature of 350°C and a maximum pressure of 300 psi.