Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 116-9e 10mM 1mL | 831217-43-7 | 1 ML
SDP

Catalog No. 5000226193
Encompass_Preferred

Small-molecule inhibitor of the Hsp70 co-chaperone DNAJA1 that inhibits SV40 replication and suppresses tumor antigen (TAg) ATPase activity. Supplied as a 10 mM solution in DMSO, 1 mL.

  • Inhibits Hsp70 co-chaperone activity and viral DNA replication.
  • Provided as a 10 mM solution in DMSO (1 mL) for ready use.
  • High purity (98.28%) suitable for biochemical assays.
  • Molecular weight 512.60; formula C31H32N2O5.
  • Good solubility in DMSO (100 mg/mL reported).
  • Recommended storage: powder and solution temperature limits to preserve stability.

Catalog No. 50-002-26193 Supplier Medchemexpress LLC Supplier No. HY11668310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Small-molecule inhibitor of the Hsp70 co-chaperone DNAJA1 that inhibits SV40 replication and suppresses tumor antigen (TAg) ATPase activity. Supplied as a 10 mM solution in DMSO, 1 mL.

  • Inhibits Hsp70 co-chaperone activity and viral DNA replication.
  • Provided as a 10 mM solution in DMSO (1 mL) for ready use.
  • High purity (98.28%) suitable for biochemical assays.
  • Molecular weight 512.60; formula C31H32N2O5.
  • Good solubility in DMSO (100 mg/mL reported).
  • Recommended storage: powder and solution temperature limits to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.