Learn More
TARGETMOL CHEMICALS INC 116-9e 5MG
SDP
Catalog No. 502877506
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.116-9e (MAL2-11B) is a potent Hsp70 co-chaperone DNAJA1 inhibitor with antiviral properties, inhibiting simian virus 40 (SV40) replication, tumour antigen (TAg) endogenous ATPase activity, and TAg-mediated activation of Hsp70. 116-9e can be used to study cancer drug resistance. Purity 99.55%
Catalog No. 50-287-7506 Supplier TARGETMOL CHEMICALS INC Supplier No. T257705MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.116-9e (MAL2-11B) is a potent Hsp70 co-chaperone DNAJA1 inhibitor with antiviral properties, inhibiting simian virus 40 (SV40) replication, tumour antigen (TAg) endogenous ATPase activity, and TAg-mediated activation of Hsp70. 116-9e can be used to study cancer drug resistance. Purity 99.55%