Learn More
Medchemexpress LLC 116-9e | 831217-43-7 | 98.3% | 512.60 | 5 MG
SDP
Catalog No. 5000364236
Encompass_Preferred
116-9e (MAL2-11B) is a Hsp70 co-chaperone DNAJA1 inhibitor. It inhibits Simian Virus 40 (SV40) replication and DNA synthesis. This product also inhibits tumor antigen (TAg)'s endogenous ATPase activity and the TAg-mediated activation of Hsp70.
  • Inhibits TAg stimulation of Hsp70 with greater efficacy than MAL3-101.
  • Significantly reduces viral replication and DNA synthesis.
  • Inhibits the activity of the TAg ATPase domain.
  • Significantly reduces the growth of BK virus in a human kidney cell line.
Catalog No. 50-003-64236 Supplier Medchemexpress LLC Supplier No. 5MGHY116683
116-9e (MAL2-11B) is a Hsp70 co-chaperone DNAJA1 inhibitor. It inhibits Simian Virus 40 (SV40) replication and DNA synthesis. This product also inhibits tumor antigen (TAg)'s endogenous ATPase activity and the TAg-mediated activation of Hsp70.
  • Inhibits TAg stimulation of Hsp70 with greater efficacy than MAL3-101.
  • Significantly reduces viral replication and DNA synthesis.
  • Inhibits the activity of the TAg ATPase domain.
  • Significantly reduces the growth of BK virus in a human kidney cell line.