Learn More
Perkin Elmer US LLC 1/16 in., 6-Port External Sample Injector
Supplier:  Perkin Elmer US LLC N9302814
Encompass_Preferred
This external sample injector features 6 ports and has a fitting of 1/16 inches. Maximum temperature of 350°. This switching valve allows employment of different desired mobile phases by periodically injecting into the carrier gas stream of the chromatograph.
Catalog No. 50-210-4018