Learn More
Sino Biological Recombinant Human AKR1A1 Protein (His Tag)
Supplier:  Sino Biological 11607H07E100
Encompass_Preferred
A DNA sequence encoding the human AKR1A1 (P14550) (Met 1-Tyr 325) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7028