Learn More
Sino Biological NRG1 Beta 1 Protein, Human, Recombinant (ECD)
Supplier:  Sino Biological 11609HNCH1
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment (Ser 2-Lys 246) of human NRG1 isoform beta1 (Q02297-6) was expressed.
Catalog No. 50-161-7038