Learn More
Sino Biological Recombinant Human SIRP alpha/SHPS-1 Protein (mFc Tag), HPLC-verified
Supplier:  Sino Biological 11612H38H100
Encompass_Preferred
A DNA sequence encoding the human SIRPA (NP_542970.1) (Met1-Arg370) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-161-7052