Learn More
Sino Biological Recombinant Human Neuroligin 1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11617H08H100
Encompass_Preferred
A DNA sequence encoding the human NLGN1 (NP_055747.1) extracellular domain (Met 1-Ser 677) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7057