Learn More
Sino Biological Recombinant Human PCBP1 Protein (His Tag)
Supplier:  Sino Biological 11628H07E100
Encompass_Preferred
A DNA sequence encoding the human PCBP1 (NP_006187.2) (Asp2-Ser356) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7067