Learn More
Sino Biological Recombinant Human RAGE Protein (hFc Tag)
Supplier:  Sino Biological 11629H02H100
Encompass_Preferred
A DNA sequence encoding the human AGER isoform 1 (NP_001127.1) extracellular domain (Met 1-Ala 344) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7069