Learn More
Sino Biological Recombinant Human RAGE Protein, HPLC-verified
Supplier:  Sino Biological 11629HCCH100
Encompass_Preferred
A DNA sequence encoding the mature form of human AGER (NP_001127.1) (Met1-Ala 344) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-161-7073