Learn More
Sino Biological Recombinant Human DLL1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11635H08H100
Encompass_Preferred
A DNA sequence encoding the human DLL1 (NP_005609.3) extracellular domain (Met 1-Gly 540) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7077