Learn More
Sino Biological Recombinant Human ALDH1A3 Protein (His Tag)
Supplier:  Sino Biological 11636H07E50
Encompass_Preferred
A DNA sequence encoding the human ALDH1A3 (NP_000684.2) (Ala 2-Pro 512) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7080