Learn More
Sino Biological Recombinant Human REG1B Protein (His Tag)
Supplier:  Sino Biological 11638H08H100
Encompass_Preferred
A DNA sequence encoding the human REG1B (NP_006498.1) (Met 1-Asn 166) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7081