Learn More
Sino Biological Recombinant Human Lumican Protein (His Tag)
Supplier:  Sino Biological 11640H08H100
Encompass_Preferred
A DNA sequence encoding the human LUM (NP_002336.1) (Met 1-Asn 338) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7085