Learn More
Sino Biological Recombinant Human Adrenomedullin Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 11646H01H50
Encompass_Preferred
A DNA sequence encoding the human ADM (P35318) (Tyr95-Tyr146) was expressed, with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-7090