Learn More
Sino Biological Recombinant Human Jagged 1/JAG1 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 11648H02H100
Encompass_Preferred
A DNA sequence encoding the human JAG1 (NP_000205.1) extracellular domain (Met 1-Ser 1046) was fused with Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7093