Learn More
Sino Biological Recombinant Human AIM2 Protein (GST Tag)
Supplier:  Sino Biological 11654H09B50
Encompass_Preferred
A DNA sequence encoding the human AIM2 (NP_004824.1) (Met 1-Thr 343) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-7102