Learn More
Sino Biological Recombinant Human TLT-2/TREML2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11655H08H100
Encompass_Preferred
A DNA sequence encoding the human TREML2 (NP_079083.2) extracellular domain (Met 1-Ser 268) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7103