Learn More
Sino Biological Recombinant Human EphA7 Protein (His & GST Tag)
Supplier:  Sino Biological 11657H20B150
Encompass_Preferred
A DNA sequence encoding the human EPHA7 (NP_004431) (Gly579-Val998) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7108